Accredo Safety Pin And Sleeve - Plug Lockout, ZING Enterprises
Supplier: ZING Enterprises
7268
76026-374EA 61.48 CAD
76026-374
Accredo Safety Pin And Sleeve - Plug Lockout, ZING Enterprises
Security Locks
Supports OSHA Standard 1910.147.
  • High quality
  • Durable
  • Lockout tagout device
Pin and Sleeve Plug Lockout prevents plugs from being placed into sockets. The device features durable nylon construction and measures 1.5"H x 2.75"L x 1.75"W. The lockout accommodates 1 padlock, with a maximum shackle diameter of ¼’’. The device is for use with 500V plugs.
Certifications: OSHA 1910.147