EZwaste® HD 83 mm Cap Assembly for HPLC Solvent Waste, Carbon Air Filter, Foxx Life Sciences
Supplier: VWR International
10827-856 10827-858 10827-860 10827-862 10827-864 76018-458 76018-460 76018-462 76018-464 76018-470 76018-472 76018-506 76018-508 76018-510
10827-856EA 870.75 CAD
10827-856 10827-858 10827-860 10827-862 10827-864 76018-458 76018-460 76018-462 76018-464 76018-470 76018-472 76018-506 76018-508 76018-510
EZwaste® HD 83 mm Cap Assembly for HPLC Solvent Waste, Carbon Air Filter, Foxx Life Sciences
Closures Bottle Closures
This cap system allows for the safe and effective collection of waste in the laboratory
  • Affordable solution for HPLC liquid waste disposal
  • VersaCap® technology
  • Carbon exhaust filter traps VOCs
  • Variety of O.D. tubing port configurations
  • Hose Barb option in 1/4" or 3/8"
The EZwaste® Solvent Waste System is designed to be an affordable solution forcontaining liquid chromatography waste. Each system is manufactured to safely collect waste from one or more HPLC systems while the carbon exhaust filters improve air quality by capturing potentially harmfulMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable