Isolator Tool™ and Covers, Micronova
Supplier: Micronova
ITC6-9 ITC6-9IR QDIT-09 ITC620-911IR QPIT-09 ITC620-911 ITC11-9NS ITC11-9NSIR ITCAW-9 ITCAA-9IR ITC6-9NS ITC6-9NSIR ITC6-911NSIR ITC11-9NSBB QPIT-04 ITC6-4NS
89040-914CS 808.14 CAD
89040-914 89040-920 89040-926 89089-356 89092-312 89140-698 94023-774 94023-776 89402-988 52428-592 52428-594 89508-392 89508-394 76018-368 76018-370 76018-372 76018-374 76018-376 76018-378 76018-380 76443-130 76443-132
Isolator Tool™ and Covers, Micronova
Mops
The Isolator Tool™ adapter with a 360° swivel works with a range of mop covers to efficiently clean and disinfect isolators, hoods, RABS, and small equipment in clean and aseptic areas.
  • Ergonomic cleaning for isolators, hoods, RABS, and other small equipment
  • Low-profile design and angled head adapter
  • Modular and extendable handles available
  • Available with stainless steel or plastic plates
  • For use with the SSU series Universal Handles or STAH series threaded handles
  • PolySorb™ Cover: 100% textured polyester for additional cleaning as well as sanitization
More Product Information