Particle Analysis Flow Cell (Lightpath: 2 mm), Type 77
Supplier: FIREFLYSCI, INC.
77IR2
76016-334EA 2900.11 CAD
76016-334
Particle Analysis Flow Cell (Lightpath: 2 mm), Type 77
Cuvettes Spectrophotometer Cuvettes
2 mm flow through cell that can be used for flow cytometry, particle analysis, liquid chromatography, and many other applications.
  • Fire-fused to ensure that cuvettes are robust and durable
  • Available in UV quartz and IR quartz
  • Comes with standard 2 mm lightpath, but can be modified if needed
  • All surfaces polished
  • Contains square flow through channel
This design can be modified for specific use (contact an Avantor sales representative for more information).