Macro Cuvettes, Type 11, FireFlySci
Supplier: FIREFLYSCI, INC.
11G1 11G2 11G5 11G10 11G20 11G40 11G50 11UV1 11UV2 11UV5 11UV10 11UV20 11UV40 11UV50 11IR1 11IR2 11IR5 11IR10 11IR20 11IR40 11IR50
76015-328EA 1027.33 CAD
76015-328 76015-330 76015-332 76015-334 76015-336 76015-338 76015-340 76015-342 76015-344 76015-346 76015-348 76015-350 76015-352 76015-354 76015-356 76015-358 76015-360 76015-362 76015-364 76015-366 76015-368
Macro Cuvettes, Type 11, FireFlySci
Cuvettes Spectrophotometer Cuvettes
FireflySci's Type 11 cuvette comes standard with a glass cap, which provides an airtight seal, ensured by the ground finish on the inside of the cap and the top of the cuvette.
  • Hand made ensuring the highest quality optical polish with no defects
  • Firefly cuvettes are guaranteed to give high optical transmission
  • Fire fused to ensure the best bonding
  • Several different materials available: (Optical Glass, UV Quartz, IR Quartz)
  • Lightpaths from 1 mm up to 50 mm available
  • Two windows optically polished
The cap is polished, which enables optical transmission.More Product Information