Semi-Micro Cuvettes, with PTFE Stopper, Firefly
Supplier: FIREFLYSCI, INC.
29UV5 29UV10 29UV20 29UV30 29UV40 29UV50 29IR5 29IR10 29IR20 29IR30 29IR40 29IR50
76015-224EA 648.12 CAD
76015-224 76015-754 76015-756 76015-758 76015-760 76015-762 76015-764 76015-802 76015-804 76015-806 76015-808 76015-810
Semi-Micro Cuvettes, with PTFE Stopper, Firefly
Cuvettes Spectrophotometer Cuvettes
Cuvettes are made with different materials like UV quartz, IR quartz ensuring high quality optical polishing with no defects.
  • Highly accurate results
  • Fire-fused to ensure that cuvettes are robust and durable
  • Base thickness is 3 mm
  • Two windows optically polished
Order Now
SPECIFICATIONS
Environmentally Preferable