Micro Fluorescence Cuvettes with PTFE Cover, FireflySci
Supplier: FIREFLYSCI, INC.
4G5X5X33 4G5X5X45 4UV5X5X33 4UV5X5X45 4IR5X5X33 4IR5X5X45
76015-184EA 409.14 CAD
76015-184 76015-186 76015-188 76015-190 76015-192 76015-194
Micro Fluorescence Cuvettes with PTFE Cover, FireflySci
Cuvettes Spectrophotometer Cuvettes
High-quality micro cuvettes in a variety of material types and are a great value.
  • Fluorescence cells with micro capacities (0.350 to 0.700 ml)
  • Comes in several different materials including optical glass, UV quartz and IR quartz
  • Lightpaths of 5 mm available with heights of 33 and 45 mm
  • Four windows and base optically polished
  • Includes PTFE cover
Firefly's micro fluorometer cuvette with PTFE cover. Made by hand, ensuring high quality optical polishing with no defects. Each cell goes through a vigorous inspection process before leaving the factory. Fire-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable