VWR® 3D Cell Culture Dishes and Plates
Supplier: VWR International
76012-952 76012-954 76012-956 76012-958 76012-960 76012-962
76012-952CS 1333.32 CAD
76012-952 76012-954 76012-956 76012-958 76012-960 76012-962
VWR® 3D Cell Culture Dishes and Plates
Culture Dishes Cell Culture Dishes
The 3D Scaffold for cell culture is able to simulate the three-dimensional structure of mamalian cells
  • 3D Scaffold is made from virgin polystyrene with a wire diameter of 500 µm
  • Scaffold spacing is a highly consistent 260 µm
  • Structured with 3-dimensional channels, offering extremely high connectivity to facilitate nutrient transmission
  • Consistency of metabolic activity and the accuracy of culture results in 3D cell culture
  • Cytokine and growth factor resistant
  • Easy cell secretion collection
Open pores with high connectivity, facilitating nutrientMore Product Information