VWR® Vacuum Filtration Systems, Standard Line
Supplier: VWR International
10040-436 10040-438 10040-440 10040-442 10040-444 10040-446 10040-448 10040-450 10040-460 10040-462 10040-464 10040-466 10040-468 10040-470 10040-472 10040-474 76008-572 76008-574 76008-576 76008-578 76008-588 76008-590 76008-592 76008-594 76008-596 76008-598 76008-600 76008-602 76008-604 76008-606 76008-608 76008-610 76008-612 76008-614 76008-616 76008-618 76008-620 76008-622 76008-624 76008-626 76008-708 76008-710 76010-368 76010-370 76010-372 76010-374 76010-376 76010-378 76010-380 76010-382 76010-384 76010-386 76010-388 76010-390 76010-392 76010-394 76010-396 76010-398 76010-400 76010-402 76010-404 76010-406 76010-408 76010-410 76010-412 76010-414 76010-416 76010-418 76010-420 76010-422 76012-768 76012-770
10040-436CS 250.1 CAD
10040-436 10040-438 10040-440 10040-442 10040-444 10040-446 10040-448 10040-450 10040-460 10040-462 10040-464 10040-466 10040-468 10040-470 10040-472 10040-474 76008-572 76008-574 76008-576 76008-578 76008-588 76008-590 76008-592 76008-594 76008-596 76008-598 76008-600 76008-602 76008-604 76008-606 76008-608 76008-610 76008-612 76008-614 76008-616 76008-618 76008-620 76008-622 76008-624 76008-626 76008-708 76008-710 76010-368 76010-370 76010-372 76010-374 76010-376 76010-378 76010-380 76010-382 76010-384 76010-386 76010-388 76010-390 76010-392 76010-394 76010-396 76010-398 76010-400 76010-402 76010-404 76010-406 76010-408 76010-410 76010-412 76010-414 76010-416 76010-418 76010-420 76010-422 76012-768 76012-770
VWR® Vacuum Filtration Systems, Standard Line
Filters Bottle Top Filters
Systems for vacuum filtration of aqueous solutions including cell culture media, buffers, or other biological fluids.
  • Polystyrene funnel and collection reservoir
  • Each individual unit is lot numbered for traceability
  • Solution can be stored in the collection reservoir until needed
  • Top filter and bottom collector bottle have engraved graduation
  • Designed hose connector can fit multiple hose diameters
PES/PVDF/Nylon/SFCA membrane: Intended for the sterile vacuum filtration of aqueous solutions such as cell culture media and biological fluids.