ZR-96 MagStand, Zymo Research
Supplier: Zymo Research
P1005
76005-120EA 892.34 CAD
76005-120
ZR-96 MagStand, Zymo Research
Beads Magnetic Beads
The ZR-96 MagStand is a magnetic stand for the separation of magnetic beads in standard 96-well format plates, especially for EZ-96 DNA Methylation MagPrep product line, ZR-96 Genomic DNA MagPrep, Zyppy-96 Plasmid MagBead Miniprep, and Direct-zol-96 MagBead RNA.