VWR® Measuring Cylinder, Hexagonal Base, TC, with Spout, Class A, Unserialized
Supplier: VWR International
76003-672 76003-674 76003-676 76003-678 76003-680 76004-046 76004-048 76004-050
76003-672CS 332.85 CAD
76003-672 76003-674 76003-676 76003-678 76003-680 76004-046 76004-048 76004-050
VWR® Measuring Cylinder, Hexagonal Base, TC, with Spout, Class A, Unserialized
Cylinders Measuring Cylinders
VWR® Measuring Cylinders are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Hexagonal base
  • Calibrated 'to Contain'
  • Cylinders are made to meet ASTM E 1272 norms
  • Printed double scale
VWR® Measuring Cylinders are used for the precise measurement liquid volumes. Cylinders have a large hexagonal base to increase stability. Calibration and tolerances are in compliance to standards: ASTM E1272, ASTM E542, and ASTM E694.
Certifications: Cylinders are supplied with Batch certificates.
Order Now
SPECIFICATIONS
Environmentally Preferable