FreeZone® 18 L Console Freeze Dryers with Stoppering Tray Dryer, Labconco®
Supplier: Labconco
701821010 701821015 701821030 701821040 701822050 701822070 701821110 701821115 701821130 701821140 701822150 701822170 701821150 701821170 701822110 701822115 701822130 701822140 701821050 701821070 701822010 701822015 701822030 701822040
76000-322EA 112875.2 CAD
76000-322 76000-324 76000-326 76000-328 76000-330 76000-332 76000-334 76000-336 76000-338 76000-340 76000-342 76000-344 76000-612 76000-614 76000-616 76000-618 76000-620 76000-622 76000-702 76000-704 76000-706 76000-708 76000-710 76000-712
FreeZone® 18 L Console Freeze Dryers with Stoppering Tray Dryer, Labconco®
Freeze Dryers
FreeZone® 18 Liter Console Freeze Dryers with Stoppering Tray Dryers are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum.
  • Stay close to your samples as Lyo-Works™ operating system, provides mobile connectivity
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ End Point Detection option takes the guess work out of sample dryness
New Lyo-Works™ OS providesMore Product Information