FreeZone® 18 L Console Freeze Dryers, Labconco®
Supplier: Labconco
701811050 701811070 701812010 701812015 701812030 701812040 701812050 701812070 701811110 701811115 701812110 701812115 701812130 701812140 701812150 701812170 701811210 701811215 701811230 701811240 701811250 701811270 701812210 701812215 701812230 701812240 701812250 701812270 701811010 701811015 701811030 701811040 701811130 701811140 701811150 701811170
76000-138EA 51239.4 CAD
76000-138 76000-140 76000-142 76000-144 76000-146 76000-148 76000-150 76000-152 76000-154 76000-156 76000-158 76000-160 76000-162 76000-164 76000-166 76000-168 76000-170 76000-172 76000-174 76000-176 76000-178 76000-180 76000-182 76000-184 76000-186 76000-188 76000-190 76000-192 76000-552 76000-554 76000-556 76000-558 76000-560 76000-562 76000-564 76000-566
FreeZone® 18 L Console Freeze Dryers, Labconco®
Freeze Dryers
FreeZone® 18 Liter Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches.
  • Stay close to your samples as Lyo-Works™ operating system, provides mobile connectivity
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ End Point Detection option takes the guess work out of sample dryness
New Lyo-Works™ OS provides flexibility and convenience in an easy-to-More Product Information