FreeZone® 12L Console Freeze Dryers with Stoppering Tray Dryer, Labconco®
Supplier: Labconco
701221010 701221015 701221030 701221040 701221050 701221070 701221130 701221140 701222150 701222170 711221130 711221140 711221150 711221170 711222110 711222115 711222130 711222140 711222150 711222170 701221110 701221115 701221150 701221170 701222110 701222115 701222130 711221040 711221050 711221070 711222010 711222015 711222030 711222040 711222050 711222070 711221110 711221115 701222010 701222015 701222030 701222040 701222050 701222070 701222140 711221010 711221015 711221030
76000-282EA 101807.3 CAD
76000-282 76000-284 76000-286 76000-288 76000-290 76000-292 76000-294 76000-296 76000-298 76000-300 76000-302 76000-304 76000-306 76000-308 76000-310 76000-312 76000-314 76000-316 76000-318 76000-320 76000-576 76000-578 76000-580 76000-582 76000-584 76000-586 76000-588 76000-590 76000-592 76000-594 76000-596 76000-598 76000-600 76000-602 76000-604 76000-606 76000-608 76000-610 76000-690 76000-692 76000-694 76000-696 76000-698 76000-700 76000-734 76000-736 76000-738 76000-740
FreeZone® 12L Console Freeze Dryers with Stoppering Tray Dryer, Labconco®
Freeze Dryers
FreeZone 12 Liter Console Freeze Dryers with Stoppering Tray Dryers are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum. Collectors are available to -50° C or -84° C.
  • Stay close to your samples as Lyo-Works™ operating system, provides mobile connectivity
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ End Point Detection option takes the guess work out of sample
More Product Information