FreeZone® 6 L Console Freeze Dryers with Stoppering Tray Dryer, Labconco®
Supplier: Labconco
700621000 700621010 700621015 700621030 700621070 700622000 700622010 700622015 700621140 700621150 700621170 700622100 700622110 700622115 700622130 700622140 700622150 700622170 710621000 710621010 710621015 710621030 710621040 710621050 710621070 710622000 710622030 710622040 710622050 710622070 710621100 710621110 710621115 710621130 710621140 710621150 710621170 710622100 710622110 710622115 710622130 710622140 710622150 710622170 700621040 700621050 710622010 710622015 700622030 700622040 700622050 700622070 700621100 700621110 700621115 700621130
76000-194EA 90723.6 CAD
76000-194 76000-196 76000-198 76000-200 76000-202 76000-204 76000-206 76000-208 76000-210 76000-212 76000-214 76000-216 76000-218 76000-220 76000-222 76000-224 76000-226 76000-228 76000-230 76000-232 76000-234 76000-236 76000-238 76000-240 76000-242 76000-244 76000-246 76000-248 76000-250 76000-252 76000-254 76000-256 76000-258 76000-260 76000-262 76000-264 76000-266 76000-268 76000-270 76000-272 76000-274 76000-276 76000-278 76000-280 76000-568 76000-570 76000-572 76000-574 76000-672 76000-674 76000-676 76000-678 76000-680 76000-684 76000-686 76000-688
FreeZone® 6 L Console Freeze Dryers with Stoppering Tray Dryer, Labconco®
Freeze Dryers
FreeZone 6 Liter Console Freeze Dryers with Stoppering Tray Dryers are designed for lyophilizing moderate to large loads and stoppering vials under vacuum. Collectors are available to -50° C or -84° C.
  • Stay close to your samples as Lyo-Works™ operating system, provides mobile connectivity
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • End-Zone™ End Point Detection option takes the guess work out of sample dryness