BIO-SILICO® Silicone Stoppers, Hirschmann
Supplier: Hirschmann
8905912 8905915 8905917 8905919 8905922 8905924 8905928 8905932 8905942 8905952
75996-694PK 151.27 CAD
75996-694 75996-696 75996-698 75996-700 75996-702 75996-704 75996-706 75996-708 75996-710 75996-712
BIO-SILICO® Silicone Stoppers, Hirschmann
Stoppers Silicone Stoppers
BIO-SILICO® stoppers are manufactured in a special process which have regular pores, thus making them ideal for the preparation and sterilization of culture media
  • High degree of air permeability, similar to that of cotton plugs
  • Autoclavable
  • Resistant to chemicals
  • Water repellent and reusable
The uniform pores and good permeability to air make the stoppers ideally suitable for cultivation of aerobic microorganisms.
Order Now
SPECIFICATIONS
Environmentally Preferable