Methanizer (CH4izer) for Agilent GC-FID₂, Restek
Supplier: Restek
22650-AU 22650-CN 22650-EU 22650-UK 22650-US
75996-502EA 4742.37 CAD
75996-502 75996-632 75996-634 75996-636 75996-638
Methanizer (CH4izer) for Agilent GC-FID₂, Restek
Gas Detection Systems
Methanizer allows ppb-level GC-FID determination of CO and CO2 without expensive equipment.
  • Simple, reliable operation
  • Precise temperature control
  • Easy tube replacement
Precise temperature control ensures complete conversion of CO and CO2 to CH4
Low-level detection of carbon monoxide (CO) and carbon dioxide (CO2) is critical for many applications. Restek’s methanizer (CH4izer) allows ppb-level determination of CO and CO2 using a cost-effective GC-FID instead of more expensive instrumentation. The methanizer is factory set to 380 °C to ensureMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable