Freer Elevator
Supplier: Sklar
98-748
75996-286EA 47.31 CAD
75996-286
Freer Elevator
Retractors
Double-ended Freer elevator is an orthopedic instrument that can be used as a multipurpose tool in many procedures, although it is primarily used for blunt debulking and lifting periosteum from bones in confined areas.
  • Multipurpose
  • Reusable
  • Nonsterile
This double-ended instrument typically has a rounded handle with two slightly curved, tear-drop tips.
Merit™ instruments are Sklar’s office-grade line of products and most commonly utilized for routine and in-office minor procedures. These cost effective, high quality instruments are crafted fromMore Product Information