Green Thermoset Screw Caps, F217 and PTFE Liner, Qorpak®
Supplier: Qorpak
CAP-00066 CAP-00068 CAP-00073 CAP-00079 CAP-00061 CAP-00063 272626 272628 272630 CAP-00065 CAP-00075 272487 272490 272491 272492 272494 272497 CAP-00060 CAP-00067 CAP-00069 CAP-00071 CAP-00072 CAP-00074 CAP-00078
16161-337CS 3874.27 CAD
16161-337 16161-360 16161-428 16161-519 16161-541 16161-574 75993-606 75993-612 75993-620 76175-390 76313-928 76539-280 76539-282 76539-284 76539-286 76539-288 76539-290 76539-292 76539-294 76539-296 76539-298 76539-300 76539-302 76539-304
Green Thermoset Screw Caps, F217 and PTFE Liner, Qorpak®
Closures Bottle Closures
Green Thermoset Plastic Caps with F217 and PTFE liners are known for providing the widest range of chemical compatibility.
  • The color green easily identifies the F217 & PTFE liner system
  • F217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature
  • Provides odor and impact resilience for a tight seal with a low moisture transmission rate
The F217 features a low-density foam core between two LDPE discs. They are applied to the back of a PTFE liner.
Some uses for these F217/PTFE lined caps include analytical labMore Product Information