VWR® Baffled Erlenmeyer Flask with GL 45 Screw Cap
Supplier: VWR International
75993-572 75993-574 75993-576 75993-578
75993-572CS 311.51 CAD
75993-572 75993-574 75993-576 75993-578
VWR® Baffled Erlenmeyer Flask with GL 45 Screw Cap
Flasks Erlenmeyer Flasks
These baffled flasks fit standard shaking incubator clamps and can be placed on any platform.
Tolerance: 2 mm
Ordering information: Flasks are supplied with polypropylene GL45 thread caps.