Corning® LSE™ Digital Dry Bath Heaters, 230 V
Supplier: Corning
6886-DB 6887-DB 6896-FB 6897-FB 6876-SB 6877-SB
75989-420CS 1072.15 CAD
75989-420 75989-422 75989-428 75989-430 75990-498 75990-500
Corning® LSE™ Digital Dry Bath Heaters, 230 V
Block Heaters Digital Block Heaters
The Corning Digital Dry Bath Heaters are available in single, dual, and four block configurations and provide a broad temperature range up to 150°C, which makes them useful for a variety of applications in molecular biology, histology, clinical, environmental, and industrial laboratories.
  • 230 V models, not for use in USA
  • Single, dual, and four block capacity to maximize sample processing
  • Dual display allows for easy monitoring of temperature and time
  • USB connectivity enables traceability of data
  • Molded block chamber provides excellent temperature
More Product Information