Ready Engineered Complete Pallet Shipping Solutions, Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
SPS GPS-RHS GPS-RHW GPS-RPS GPS-RPW GPS-RFS GPS-RFW GPS-RP-PL
10795-966PL 2724.99 CAD
10795-966 10841-256 10841-258 10841-260 10841-262 10841-264 10841-266 75983-124
Ready Engineered Complete Pallet Shipping Solutions, Sonoco ThermoSafe
Mailers and Shippers
Complete Pallet Shipping Solutions are designed to meet the unique transport requirements of pharmaceuticals, biologics, vaccines, and other high value life science products.
  • Pre-qualified for refrigerated, frozen, and CRT temperatures
  • Optimized size
Featuring more temperature options, fewer packing components, and easy assembly, these pallet solutions are designed to meet the unique transport requirements of pharmaceuticals, biologics, vaccines, and other high value life science products. Dimensions have been optimized for narrow-body aircraft, wide-More Product Information