Anti-TYRP1 Mouse Monoclonal Antibody [clone: TA99 TYRP1/807]

Supplier: Biotium

BNUM0808-50
75978-000EA 669.83 CAD
75978-000
Anti-TYRP1 Mouse Monoclonal Antibody [clone: TA99 TYRP1/807]
Antibodies
This antibody reacts with a 75 kDa melanocyte-specific gene product, identified as Tyrosinase-related protein-1 (TRP-1). It is involved in melanin synthesis. TRP1 is present on the melanosomal membranes of melanoma, normal melanocytes and nevi.Recent evidence suggests that TRP-1 is involved in maintaining stability of tyrosinase protein and modulating its catalytic activity. TRP-1 is also involved in maintenance of melanosome ultrastructure and affects melanocyte proliferation and cell death.

Type: Primary
Antigen: TYRP1
Clonality: Monoclonal
Clone: Clone TA99 TYRP1/807
Conjugation:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR