Anti-TYMS Mouse Monoclonal Antibody (CF647) [clone: TS106 TMS715]
Supplier: Biotium
BNC470716-100 BNC470716-500
75977-924EA 325.12 CAD
75977-924 75977-926
Anti-TYMS Mouse Monoclonal Antibody (CF647) [clone: TS106 TMS715]
Antibodies
This antibody recognizes a protein of 36 kDa, identified as Thymidylate Synthase (TS) (EC 2.1.1.45). TS converts deoxyuridine monophosphate (dUMP) to deoxythymidine monophosphate (dTMP), which is essential for DNA biosynthesis. TS is also a critical target for the fluoropyrimidines, an important group of antineoplastic drugs that are widely used in the treatment of solid tumors. Both 5-FU and fluorodeoxyuridine are converted in tumor cells to FdUMP which inactivates TS by formation of a ternary covalent complex in the presence of the folate cofactor 5,10-methylenetetrahydrofolate. ExpressionMore Product Information