Anti-MUC1 Mouse Monoclonal Antibody [clone: MUC1/955]
Supplier: Biotium
BNUB0955-100 BNUB0955-500
75963-528EA 380.65 CAD
75963-528 75963-530
Anti-MUC1 Mouse Monoclonal Antibody [clone: MUC1/955]
Antibodies
This MAb reacts with MUC1. The dominant epitope of this MAb has not yet been determined. MUC1 is a large cell surface mucin glycoprotein expressed by most glandular and ductal epithelial cells and some hematopoietic cell lineages. It is expressed on most secretory epithelium, including mammary gland and some hematopoietic cells. It is expressed abundantly in lactating mammary glands and over expressed abundantly in >90% breast carcinomas and metastases. Transgenic MUC1 has been shown to associate with all four c-erbB receptors and localize with c-erbB1 (EGFR) in lactating glands. The MUC1More Product Information