Anti-GLG1 Mouse Monoclonal Antibody (CF647) [clone: GLG1/970]
Supplier: Biotium
BNC470970-100 BNC470970-500
75927-624EA 325.12 CAD
75927-624 75927-626
Anti-GLG1 Mouse Monoclonal Antibody (CF647) [clone: GLG1/970]
Antibodies
This MAb recognizes a protein of 134 kDa, which binds fibroblast growth factor and E-selectin (cell-adhesion lectin on endothelial cells mediating the binding of neutrophils). Fucosylation is essential for binding to E-selectin. It contains sialic acid residues and 16 Cys-rich GLG1 repeats. This MAb can be used to stain the Golgi complex in cell or tissue preparations and can be used as a Golgi marker in subcellular fractions. It produces a diffuse staining pattern of the Golgi zone in normal and malignant cells. This MAb is an excellent marker for human cells in xenographic model research.More Product Information