Anti-Double Stranded DNA Mouse Monoclonal Antibody (CF488A) [clone: AE-2]
Supplier: Biotium
BNC880946-100 BNC880946-500
75924-232EA 325.12 CAD
75924-232 75924-234
Anti-Double Stranded DNA Mouse Monoclonal Antibody (CF488A) [clone: AE-2]
Antibodies
This monoclonal antibody is part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. This MAb recognizes the double stranded DNA in human cells. It can be used to stain the nuclei in cell or tissue preparations and can be used as a nuclear marker in human cells. This MAb produces a homogeneous staining pattern in the nucleus of normal and malignant cells.,Double Stranded deoxyribonucleic acid (ds DNA) is the genetic material of allMore Product Information