Anti-CEACAM1 & CEACAM5 Mouse Monoclonal Antibody [clone: C66/1030]
Supplier: Biotium
BNUM1030-50
75915-448EA 669.83 CAD
75915-448
Anti-CEACAM1 & CEACAM5 Mouse Monoclonal Antibody [clone: C66/1030]
Antibodies
This antibody recognizes proteins of 80-200 kDa, identified as different members of CEA family. CEA is synthesized during development in the fetal gut and is re-expressed in increased amounts in intestinal carcinomas and several other tumors. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEA is not found in benign glands, stroma, or malignant prostatic cells. Antibody to CEA is useful in detecting early foci ofMore Product Information