Anti-CD46 Mouse Monoclonal Antibody (CF568) [clone: 197-2B1]
Supplier: Biotium
BNC680931-100 BNC680931-500
75913-686EA 325.12 CAD
75913-686 75913-688
Anti-CD46 Mouse Monoclonal Antibody (CF568) [clone: 197-2B1]
Antibodies
This antibody recognizes a protein of 52 kDa-58 kDa, identified as CD46 (also known as membrane cofactor protein, MCP). CD46 exists as many isoforms in a variety of tissues. It is strongly expressed on salivary gland ducts and kidney ducts, moderately on lymphocytes and endothelium, and weakly on interstitial tissues and muscle cells, but not on erythrocytes. CD46 functions as a C3b/C4b-binding glycoprotein that inhibits complement activation on host cells. It also serves as a measles virus receptor, an adherence factor for group A Streptococcus pyogenes, and a cellular pilus receptor forMore Product Information