Anti-CD46 Mouse Monoclonal Antibody [clone: 197-2B1]

Supplier: Biotium

BNUB0931-100 BNUB0931-500
75913-652EA 380.65 CAD
75913-652 75913-654
Anti-CD46 Mouse Monoclonal Antibody [clone: 197-2B1]
Antibodies
This antibody recognizes a protein of 52 kDa-58 kDa, identified as CD46 (also known as membrane cofactor protein, MCP). CD46 exists as many isoforms in a variety of tissues. It is strongly expressed on salivary gland ducts and kidney ducts, moderately on lymphocytes and endothelium, and weakly on interstitial tissues and muscle cells, but not on erythrocytes. CD46 functions as a C3b/C4b-binding glycoprotein that inhibits complement activation on host cells. It also serves as a measles virus receptor, an adherence factor for group A Streptococcus pyogenes, and a cellular pilus receptor forMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD46
Environmentally Preferable
Antigen name CD46
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 197-2B1
Reactivity Human
Host Mouse
Gene ID Unigene ID: 510402
Isotype IgG2a kappa
ImmunoFluorescence Yes
Concentration 0.2 mg/mL
Storage buffer PBS/0.05% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR