Anti-CD84 Mouse Monoclonal Antibody [clone: 153-4D9]

Supplier: Biotium

BNUM1049-50
75910-778EA 669.83 CAD
75910-778
Anti-CD84 Mouse Monoclonal Antibody [clone: 153-4D9]
Antibodies
This antibody recognizes a protein of 74 kDa, identified as CD84. It is expressed on mature B cells and on B-cell lines, including pre-B-cell lines, but not on plasma cell lines. Immunohistochemical studies demonstrated that CD84 strongly expressed on tissues macrophages. CD84 is also highly expressed on platelets and, at low levels on peripheral blood T cells. It is a highly N-glycosylated protein and belongs to immunoglobulin superfamily. It may play a role in leukocyte activation.

Type: Primary
Antigen: CD84
Clonality: Monoclonal
Clone: 153-4D9
Conjugation: Unconjugated, BSA-and azide-More Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD84
Environmentally Preferable
Antigen name CD84
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 153-4D9
Reactivity Human
Host Mouse
Gene ID Unigene ID: 398093
Isotype IgG1 kappa
ImmunoFluorescence Yes
Concentration 1 mg/ml
Storage buffer PBS, no BSA, no azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR