Anti-BSG Mouse Monoclonal Antibody (CF594) [clone: BSG/963]
Supplier: Biotium
BNC940963-100 BNC940963-500
75887-758EA 325.12 CAD
75887-758 75887-760
Anti-BSG Mouse Monoclonal Antibody (CF594) [clone: BSG/963]
Antibodies
This MAb recognizes extracellular epitope of human CD147. It is expressed more intensely on thymocytes than on mature peripheral blood T cells. CD147 is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. It stimulates the production of interstitial collagenase, gelatinase A, stromelysin-1 and various metalloproteinases (MMPs) by fibroblasts. These enzymes are important factors in cancer invasion and metastasis.
CF® dyes are Biotium's next-generation fluorescent dyes. CF®594 is a deep red fluorescent dye (Ex/Em 593/614 nm). It yields theMore Product Information