Anti-AMPD3 Mouse Monoclonal Antibody [clone: AMPD3/901]

Supplier: Biotium

BNUB0901-100 BNUB0901-500
75881-640EA 380.65 CAD
75881-640 75881-642
Anti-AMPD3 Mouse Monoclonal Antibody [clone: AMPD3/901]
Antibodies
This antibody recognizes a protein of ~90 kDa, which is identified as Adenosine Monophosphate Deaminase, isoform E (AMPD3). It has 767 amino acids and is assigned an EC 3.5.4.6. It is a highly regulated enzyme that catalyzes the hydrolytic deamination of adenosine monophosphate to inosine monophosphate, a branch point in the adenylate catabolic pathway. AMPD3 gene encodes the erythrocyte (E) isoforms, whereas other family members encode isoforms that predominate in muscle (M) and liver (L) cells. This MAb shows reactivity with cells of the erythroid lineage at all stages of maturation in theMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol AMPD3
Environmentally Preferable
Antigen name Adenosine Monophosphate Deaminase 3
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone AMPD3/901
Reactivity Human
Host Mouse
Gene ID Unigene ID: 501890
Isotype IgG2b kappa
ImmunoChemistry Yes
ImmunoFluorescence Yes
Concentration 0.2 mg/mL
Storage buffer PBS/0.05% BSA/0.05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR