Excelta Corp® Pin Vises, Brass
Supplier: Excelta
227A-ET 227B 227B-ET 227C-ET 227C-P 227D-ET
63041-536EA 38.33 CAD
63041-536 63041-538 63041-540 63041-542 63043-694 89411-690 75880-876
Excelta Corp® Pin Vises, Brass
Dissection Needles
The 277 series of pin vise provides handle for various drill bits or probes.
  • Miniature design for accessibility under scope
  • Available in two different chuck sizes
  • Serrated handle for improved gripping
  • Available with ergonomic grip
The 'Series 227' style is a brass pin vice specifically designed to handle small drills or probes. The 227A has a 0" -.015" opening chuck. The 227B has a 0 to .025" opening chuck. The 227C and 227D include a .010" replaceable probe. The suffix 'ET' features the Ergo-Tweeze® soft cushioned grips (10¹⁰ ohms/sq surfaceMore Product Information