Plier, Large Frame, Excelta
Supplier: Excelta
907-ICA
75880-658EA 791.15 CAD
75880-658
Plier, Large Frame, Excelta
Pliers
These pliers form stress relief for thru-hole components.
  • Large frame
  • Long cushioned grips – ESD safe (1010 ohms/sq)
They feature dual leaf springs for smooth action. Model 907-ICA forms stress relief on multiple leads.