Scissors, Hard Wire Carbide Insert Blades, Excelta Corp®
Supplier: Excelta
279-HT 346-6-HT 347-6-HT 270-HT 271-HT
75880-574EA 143.78 CAD
75880-574 75880-594 75880-596 75880-892 75880-894
Scissors, Hard Wire Carbide Insert Blades, Excelta Corp®
Scissors
These slim stainless steel scissors has tungsten carbide insert blades for cutting tough materials.
  • Cuts small hard wires, (carbide insert)
  • Ergonomic grips, dual leaf springs
  • Available in different head sizes
Ideal for hard wire cutting applications.