Topaz GC Inlet Liners for Agilent Gas Chromatographs, Restek
Supplier: Restek
23431 23433 23301 23302 23308 23313 23315 23428 23310 23311 23312 23430 23429 23333 23280 23434 23282 23283 23306
75877-858PK 701.73 CAD
75877-858 75877-868 75877-872 75877-874 75877-878 75877-880 75877-884 75877-932 75877-934 75877-936 75878-208 75878-216 75878-228 75878-254 75878-266 75878-268 75878-278 75878-280 75878-286
Topaz GC Inlet Liners for Agilent Gas Chromatographs, Restek
Chromatography Supplies Chromatography Inlet Liners
Topaz GC inlet liners feature revolutionary technology and inertness to deliver you the next level of True Blue Performance.
  • Topaz technology deactivation
  • Low breakdown
  • Reproducible
  • Maximized GC uptime
These inlet liners feature low breakdown for accurate and precise low-level GC analyses. Liners undergo manufacturing controls and QC testing for superior reliability across compound classes, and they feature unparalleled cleanliness for maximized GC uptime and lab throughput.
Order Now
SPECIFICATIONS
Environmentally Preferable