EnzyChrom™ Oxalate Assay Kit, BioAssay Systems
Supplier: BIOASSAY SYSTEMS
EOXA-100
75878-206EA 1342.79 CAD
75878-206
EnzyChrom™ Oxalate Assay Kit, BioAssay Systems
Assays Immunological Assays
For quantitative determination of oxalate (oxalic acid) in urine and other samples.
  • Linear detection range 20 - 1500 µM oxalate
  • Sample pre-treatment is faster and easier than using activated carbon in competitor’s assay kits
  • Can be automated to test thousands of samples per day
Oxalate or Oxalic Acid is a metabolic breakdown product of the Kreb’s Cycle in eukaryotes, and the glyoxylate cycle in other microorganisms
BioAssay Systems’ oxalate assay kit uses a single Working Reagent that combines the oxalate oxidase reaction and color reaction in one step. The change in color intensity of the reaction product at 595 nm is directly proportional to oxalate in the sample.
It can be found in the urine of humans and other mammals. Oxalate concentration can be used as a measure of kidney function where a high level of oxalate is an indicator for kidney stones which are primarily made of the insoluble salt calcium oxalate. Measuring oxalate is more accurate than measuring calcium as a marker for kidney stones because calcium is excreted at high concentrations even in normal urine. Simple and high-throughput assays for measuring oxalate concentration find wide applications.