Avantor Single-Use Tank Liners, Medical Grade, Avantor Fluid Handling
Supplier: AVANTOR FLUID HANDLING, LLC
1003001 1003003 1003004 1003005 1003006 1003007 1003008 1003009 1003010 1003011 1003012
75874-384CS 842.52 CAD
75874-384 75874-388 75874-390 75874-392 75874-394 75874-396 75874-398 75874-400 75874-402 75874-404 75874-406
Avantor Single-Use Tank Liners, Medical Grade, Avantor Fluid Handling
Bags Media Bags
These tank liners are single-use bags with an open top, designed to fit in cylindrical vessels.
  • No cleaning after media preparation
  • Medical grade, multi-layer film
  • For bioprocess applications
These bags are used as a liner into a vessel to avoid cleaning of the vessel after media and buffer preparation.
Tank Liners are made of a clean medical grade multi-layer film designed for bioprocess applications. The fluid contact layer is a medical grade LLDPE. To minimize gas diffusion, an EVOH layer is coextruded between the inner and outer layers. TheMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable