SP Scientific miVac Sample Holder Jet Rotors for DNA and DUO Concentrators, Genevac
Supplier: Genevac
DRC-01075-068 DRC-01275-040 DRC-05EPP-120 DRC-12V40-078 DRC-13100-032 DRC-15CCT-012 DRC-15EPP-084 DRC-15V45-048 DRC-16100-028 DRC-16V35-048 DRC-17115-018 DRC-18150-006 DRC-18V40-048 DRC-20EPP-084 DRC-20V60-035 DRC-28V60-014 DRC-34100-008 DRC-50CCT-008
75871-508EA 2592.1 CAD
75871-508 75871-510 75871-512 75871-514 75871-516 75871-518 75871-520 75871-522 75871-524 75871-526 75871-528 75871-530 75871-532 75871-534 75871-536 75871-538 75871-540 75871-542
SP Scientific miVac Sample Holder Jet Rotors for DNA and DUO Concentrators, Genevac
Centrifuges Rotors
Lightweight, easily interchangeable sample holders for DNA and DUO Concentrators hold a variety of glass and plastic tubes and vials.
  • Lightweight solid aluminum construction
  • Friction fit on center spindle
  • Convenient handles
  • Hard-wearing and highly durable
  • Exceptional performance concentration
  • Wide selection available
Each miVac rotor is made to exacting specifications to assure long life, secure holding of samples and maximum heat transfer and reliability
Lightweight solid aluminum material has a very high thermal conductivity, allowing forMore Product Information