Forced Air Ovens, SM05, SHEL LAB
Supplier: Sheldon Manufacturing
SMO5 SMO5-2
75870-814EA 10085.41 CAD
75870-814 75870-830
Forced Air Ovens, SM05, SHEL LAB
Ovens Standard Ovens
The SMO5 ovens are recommended for critical and analytical use in laboratory and pilot plant product research. Typical applications for the oven include aging, asphalt testing, curing, drying, pre-heating, conditioning and life cycle testing.
  • Embedded microprocessor control with countdown timer
  • Horizontal forced airflow
  • 1.75" (4.5 cm) access port
  • 24 month limited warranty
The oven is equipped with a stainless steel interior for long life operation and easy cleaning
This unit has a quick ramp-up time to 306°C for faster cycle times. This unitMore Product Information