VWR® 96-Well Deep Well Microplates, Extractable-Free
Supplier: VWR International
75870-790 75870-792 75870-794 75870-796 75870-802
75870-790CS 631.43 CAD
75870-790 75870-792 75870-794 75870-796 75870-802
VWR® 96-Well Deep Well Microplates, Extractable-Free
Microplates Deep Well Microplates
A range of high-quality, dependable deep-well blocks for storage and collection, LC-MS, SPE, and mixing.
  • Guaranteed extractable-free
  • Gamma irradiated
  • Ready-to-use
  • For assured assays
  • DNase-free and RNase-free
These microplates are made of extractable-free polypropylene that will not leach unwanted compounds from the polymer.
Plates can be sealed with friction or heat seals to prevent evaporation, condensation, and contamination. These plates are ideal for sample storage and transport. Blocks are rigid and robust, and they can be easily stacked,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable