Combined pH Electrodes, Mettler
Supplier: Mettler Toledo
51109502 51109503 51109504 51109505 51109506 51109507 51109508 51109500
75870-646EA 1149.95 CAD
75870-646 75870-648 75870-650 75870-652 75870-654 75870-656 75870-674 75870-706
Combined pH Electrodes, Mettler
Electrodes pH Electrodes
Combined glass pH electrodes for direct pH measurement and acid/base titrations in aqueous or non-aqueous media.
  • Integrated glass sleeve for secure fixation in the titration head
  • With S7 screw cap and detachable cap or with Plug & Play S12 connection
  • Plug & Play sensor features an integrated sensor chip which allows data storing and monitoring
DG111-SC / DGi111-SC / DG114-SC / DGi114-SC: ARGENTHAL™ reference system
DG113-SC / DGi113-SC: Ag/AgCl/LiCl reference system
DGi101-SC / DG115-SC / DGi115-SC: ARGENTHAL™ reference system with silver ionMore Product Information