SP Bel-Art H-B Frio-Temp Calibrated Electronic Lollipop Thermometers for Refrigerators, Incubators and General Applications, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
B61000-0900 B60210-1300 B60210-1400 B60210-1500 B60210-1700 B60210-1800 B60210-1900 B60210-2300 B60210-2500 B60210-2700 B61000-0700 B61000-0800
75854-030CS 831.59 CAD
75854-030 75854-032 75854-034 75854-036 75854-038 75854-040 75854-042 75854-044 75854-046 75854-048 75854-050 75854-052 75854-054 75854-056 75854-104 75854-106
SP Bel-Art H-B Frio-Temp Calibrated Electronic Lollipop Thermometers for Refrigerators, Incubators and General Applications, Bel-Art Products, a part of SP
Thermometers Digital Thermometers
Digital verification thermometers verify sample temperatures inside refrigerators and incubators without the use of a cable.
  • Accurate temperature reading without sample contamination is achieved by using the liquid or glass bead filled bottle (included), which mimics the sample
  • All models include accredited calibration documentation that meets the requirements of ISO 17025
  • Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to
More Product Information