Dissolution Capsule Sinkers for Apparatus 2, Quality Lab Accessories
Supplier: QLA
BSK008-JP CAPWHT-02 CAPWHT-04 CAPWHT-2S CAPWHT-4S CAPWHT-LG CAPWHT-TR CAPWHT-VK CAPWHT-XL CAPWHT-XS CAPWST-15 CAPWST-18 CAPWST-23 CAPWST-31 CUSBSK-JP MAGWHT-VK SNK008-VK SNK010-VK WIRESK-VK CAPWHT-PC CAPWHT-S99 CAPWHT-SV CAPWHT-XLS CUSBSK-JPMAG SNK080-VK SNK040-VK BSK008-01 CAPWST-19 CAPWHT-XLSS BSK008-01LG CUSBSK-ALF
11010-010EA 237 CAD
11010-010 11010-138 11010-140 11010-142 11010-144 11010-146 11010-148 11010-150 11010-152 11010-154 11010-156 11010-158 11010-162 11010-164 11010-212 11010-382 11010-470 11010-472 11010-534 89009-498 89039-804 89049-234 89132-970 89132-990 89173-160 93001-040 93001-074 11010-008 11010-160 75848-862 75848-996 75849-182
Dissolution Capsule Sinkers for Apparatus 2, Quality Lab Accessories
Weights Sinkers
QLA offers the widest variety of Capsule Sinkers in the Dissolution Industry.
  • Made of high quality 316 stainless steel or PTFE coated over music wire
  • All sinkers are made in the USA
  • Custom sinkers are available upon request
Home-made sinkers tend not to be reproducible and so commercially available sinkers are quite popular
USP (711) states: " A small, loose piece of non-reactive material such as not more than a few turns of wire helix may be attached to dosage units that would otherwise float. Other validated sinker devices may be used."