E Plus Ultrasonic Bath with Pulse Mode for Mixing, Cleaning, and Degassing, Elma
Supplier: Elma
1118880-101 1118886-101 1118890-101 1118882-101 1118891-101 1118893-101 1118895-101 1118897-101 1118899-101 1118901-101
75848-740EA 1215.38 CAD
75848-740 75848-742 75848-744 75848-746 75848-748 75848-750 75848-752 75848-754 75848-756 75848-758
E Plus Ultrasonic Bath with Pulse Mode for Mixing, Cleaning, and Degassing, Elma
Ultrasonic Cleaners
The dependable, state-of-the-art 37 kHz Elmasonic E Plus series includes 9 tank sizes with simple, user-friendly operation. Samples are rapidly mixed, dispersed, dissolved, or homogenized in Dynamic mode, while Eco mode offers quiet gentle cleaning at reduced power.
  • Combine Sweep and Pulse modes in the all new Dynamic mode
  • Dynamic mode can remove stubborn contaminants, degassing, and mixing/sample prep in the lab
  • Eco mode provides a reduced power gentle cleaning, quiet operation, and extends tank lifetime
  • Elmasonic has a 2-year warranty
Dynamic modeMore Product Information