VWR® Flexible Multi-Tube Racks
Supplier: VWR International
75845-988 75845-990 75845-992 75845-994 75845-996 75845-998 75846-000 75846-072
75845-988CS 243.1 CAD
75845-988 75845-990 75845-992 75845-994 75845-996 75845-998 75846-000 75846-072
VWR® Flexible Multi-Tube Racks
Racks Tube Racks
This rack is truly adaptable, as each well adjusts to accommodate 7 to 50 mL tube sizes and holds them in a firm upright position.
  • Racks can be linked together for additional capacity
  • Open sides allow for label viewing
  • Flat base to hold tubes upright
  • Accommodates tubes from 7 to 50 mL sizes
  • Autoclavable
VWR® Flexible Multi-Tube Racks feature unique slots that adapt to accommodate varied tubes sizes and hold them in a firm upright position. Racks are manufactured from POM, and can be linked together for additional holding capacity. Linked racks shouldMore Product Information