Pig Recombinant IL-8 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
400-03AF-5UG 400-03AF-20UG 400-03AF-100UG 400-03AF-500UG 400-03AF-1MG
75845-310EA 142.55 CAD
75845-310 75845-312 75845-314 75845-316 75845-318
Pig Recombinant IL-8 (from E. coli)
Proteins and Peptides
Interleukin 8 (IL-8 or CXCL8) is a member of the CXC cytokine family and is produced by macrophages, epithelial, smooth muscle, and endothelial cells. IL-8 binds the  G protein-coupled serpentine receptors CXCR1 and CXCR2. IL-8 recruits innate immune cells, induces phagocytosis, and stimulates angiogenesis.
  • High quality
  • Low endotoxin
  • Affordable