Anti-CD13 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: WM15]

Supplier: BIOGEMS INTERNATIONAL INC.
05211-80-100
75843-908EA 951.92 CAD
75843-908
Anti-CD13 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: WM15]
Antibodies
The WM-15 monoclonal antibody specifically reacts with human CD13, 150-170 type II transmembrane ectoenzyme also named aminopeptidase N and gp150. It is expressed by granulocytes and monocytes, epithelial cells, and endothelial cells. CD13 plays a role in the terminal degradation of peptides, antigen processing, and chemokine cleavage, and cellular adhesion.

Type: Primary
Antigen: CD13
Clonality: Monoclonal
Clone: WM15
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR