Anti-F4/80 Rat Monoclonal Antibody (BG Violet 450) [clone: BM8.1]
Supplier: BIOGEMS INTERNATIONAL INC.
02922-40-100
75843-744EA 713.58 CAD
75843-744
Anti-F4/80 Rat Monoclonal Antibody (BG Violet 450) [clone: BM8.1]
Antibodies
The BM8.1 monoclonal antibody specifically binds to the Mouse125 kDa F4/80 antigen, expressed by most mature macrophages. F4/80 is a transmembrane protein used as a marker of macrophages, although it is also expressed on Kupffer and Langerhans cells. The expression of F4/80 antigen is upregulated on bone marrow cells stimulated in vitro with the macrophage colony stimulating factor. The F4/80 antigen is a requirement for the induction of CD8 T cells-mediated peripheral tolerance.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excitedMore Product Information